Drukuj

Anti-IgG F(ab')2 Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))

Dostawca: Rockland Immunochemicals
ROCK605-4504EA 2090 PLN
ROCK605-4504
Anti-IgG F(ab')2 Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))
Przeciwciała
Secondary Rabbit Anti-IgG F(ab')2 Reacts with Goat

Recommended Dilutions: ELISA: 1:6000; Western Blot: 1:500 - 1:2500; Immunochemistry: 1:200 - 1:1000;
Do not freeze!

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: AP (Alkaline Phosphatase)
Epitope: F(ab')2
Host: Rabbit
Isotype: IgG
Reactivity: Goat
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR