Drukuj

Anti-IgG (H+L) Rabbit Polyclonal Antibody

Dostawca: Rockland Immunochemicals
ROCK605-4113EA 1360 PLN
ROCK605-4113
Anti-IgG (H+L) Rabbit Polyclonal Antibody
Przeciwciała
Secondary Rabbit Anti-IgG (H&L) Reacts with Goat

Recommended Dilutions: ELISA: 1:20000 - 1:100000; Western Blot: 1:2000 - 1:10000; Immunochemistry: 1:1000 - 1:5000;

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Heavy and Light chain
Host: Rabbit
Isotype: IgG
Reactivity: Goat
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR