Drukuj

Anti-IgG F(ab')2 Rabbit Polyclonal Antibody (TRITC (Tetramethylrhodamine Isothiocyanate))

Dostawca: Rockland Immunochemicals
ROCK605-4004EA 1510 PLN
ROCK605-4004
Anti-IgG F(ab')2 Rabbit Polyclonal Antibody (TRITC (Tetramethylrhodamine Isothiocyanate))
Przeciwciała
Secondary Rabbit Anti-IgG F(ab')2 Reacts with Goat

Recommended Dilutions: FLISA: 1:10000 - 1:50000; Immunofluorescence: 1:1000 - 1:5000; Flow Cytometry: 1:500 - 1:2500
Restore with deionized water (or equivalent); 1.0 ml

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: TRITC (Tetramethylrhodamine Isothiocyanate)
Epitope: F(ab')2
Host: Rabbit
Isotype: IgG
Reactivity: Goat
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR