Drukuj

Anti-RLN2 Rabbit Polyclonal Antibody

Dostawca: Oncogene
553850-100EA 2880 PLN
553850-100
Anti-RLN2 Rabbit Polyclonal Antibody
Przeciwciała
Type: Primary
Antigen: RLN2 (relaxin 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR