Drukuj
Anti-G i alpha-1 and Rabbit Polyclonal Antibody
Dostawca: Oncogene
371723-50EA 3420 PLN
371723-50
Anti-G i alpha-1 and Rabbit Polyclonal Antibody
Przeciwciała
Type: Primary
Antigen: G i alpha-1 and
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Zobacz ceny