Drukuj

Anti-ANGPT2 Rabbit Polyclonal Antibody

Dostawca: Oncogene
176002-50EA 2880 PLN
176002-50
Anti-ANGPT2 Rabbit Polyclonal Antibody
Przeciwciała
Type: Primary
Antigen: ANGPT2 (angiopoietin 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Mouse
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR