Drukuj
Anti-IgG Goat Polyclonal Antibody (CF™ 640R)
Dostawca: Biotium
20176-1 20176
20176-1EA 734 PLN
20176-1 20176.
Anti-IgG Goat Polyclonal Antibody (CF™ 640R)
Przeciwciała
Type: Secondary
Antigen: IgG (Rabbit) (rabbit IgG)
Clonality: polyclonal
Clone:
Conjugation: CF® 640R Dye
Epitope:
Host: Goat
Isotype: none
Reactivity: Rabbit