Drukuj

Anti-IgG Goat Polyclonal Antibody (CF™ 633)

Dostawca: Biotium
20133 20133-1
20133.EA 1830 PLN
20133. 20133-1
Anti-IgG Goat Polyclonal Antibody (CF™ 633)
Przeciwciała
Type: Secondary
Antigen: IgG (Rat) (rat IgG)
Clonality: polyclonal
Clone:
Conjugation: CF® 633 Dye
Epitope:
Host: Goat
Isotype: none
Reactivity: Rat
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR