Drukuj

Anti-IgG Goat Polyclonal Antibody (CF™ 660R)

Dostawca: Biotium
20054 20054-1
20054.EA 1530 PLN
20054. 20054-1
Anti-IgG Goat Polyclonal Antibody (CF™ 660R)
Przeciwciała
Type: Secondary
Antigen: IgG (Mouse) (mouse IgG)
Clonality: polyclonal
Clone:
Conjugation: CF® 660R Dye
Epitope:
Host: Goat
Isotype: none
Reactivity: Mouse
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR