Drukuj

Anti-IgG Goat Polyclonal Antibody (CF™ 647)

Dostawca: Biotium
20040 20040-1
20040.EA 1530 PLN
20040. 20040-1
Anti-IgG Goat Polyclonal Antibody (CF™ 647)
Przeciwciała
Type: Secondary
Antigen: IgG (Mouse) (mouse IgG)
Clonality: polyclonal
Clone:
Conjugation: CF® 647 Dye
Epitope:
Host: Goat
Isotype: none
Reactivity: Mouse
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR