Drukuj

Anti-IgG Goat Polyclonal Antibody (CF™ 555)

Dostawca: Biotium
20032 20032-1
20032.EA 1220 PLN
20032. BTIU20032-1
Anti-IgG Goat Polyclonal Antibody (CF™ 555)
Przeciwciała
This conjugate is prepared by labeling F(ab’)2 fragment of goat anti-mouse IgG (H+L) with CF™555 dye. CF™ dyes offer advantages in brightness, photostability, and specificity compared to other fluorescent dyes. Orange-red fluorescent dye CF™555 (Ex/Em 555/565 nm) is brighter than Cy®3 and an excellent replacement for Alexa Fluor® 555 for antibody conjugates.

Alexa Fluor® is a registered trademark of Thermo Fisher Scientific. Cy®3 is a registered trademark of GE Healthcare.

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: CF555
Epitope:
Host: Goat
Isotype:
Reactivity: Mouse
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR