Drukuj

Anti-CD23 Rabbit Monoclonal Antibody [clone: SP163]

Dostawca: Abcam
AB135386-100 AB135386-1 AB135386-500
ABCAAB135386-100EA 3090 PLN
ABCAAB135386-100 ABCAAB135386-1 ABCAAB135386-500
Anti-CD23 Rabbit Monoclonal Antibody [clone: SP163]
Przeciwciała
Rabbit monoclonal [SP163] to CD23.

Uwaga: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: CD23
Clonality: Monoclonal
Clone: SP163
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR