Drukuj

Anti-Methylmalonyl Coenzyme A mutase Rabbit Monoclonal Antibody [clone: EPR7739]

Dostawca: Abcam
AB133672-100 AB133672-1ML
ABCAAB133672-100EA 3420 PLN
ABCAAB133672-100 ABCAAB133672-1ML
Anti-Methylmalonyl Coenzyme A mutase Rabbit Monoclonal Antibody [clone: EPR7739]
Przeciwciała
Rabbit monoclonal [EPR7739] to Methylmalonyl Coenzyme A mutase.

Uwaga: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: Methylmalonyl Coenzyme A mutase
Clonality: Monoclonal
Clone: EPR7739
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse; Rat
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR