Drukuj

Anti-CD14 Rabbit Monoclonal Antibody [clone: EPR3652]

Dostawca: Abcam
AB133503-100 AB133503-10 AB133503-20UL AB133503-1ML
ABCAAB133503-100EA 3140 PLN
ABCAAB133503-100 ABCAAB133503-10 ABCAAB133503-20UL ABCAAB133503-1ML
Anti-CD14 Rabbit Monoclonal Antibody [clone: EPR3652]
Przeciwciała
Rabbit monoclonal [EPR3652] to CD14.

Uwaga: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: CD14
Clonality: Monoclonal
Clone: EPR3652
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR