Drukuj

Anti-Eph receptor B1/NET Rabbit Monoclonal Antibody [clone: EPR6457]

Dostawca: Abcam
AB129103-100 AB129103-1ML
ABCAAB129103-100EA 2610 PLN
ABCAAB129103-100 ABCAAB129103-1ML
Anti-Eph receptor B1/NET Rabbit Monoclonal Antibody [clone: EPR6457]
Przeciwciała
Uwaga:

All products are FOR RESEARCH USE ONLY.   PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES.  Customer is granted a limited use non-exclusive licence for its own internal research.  Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product.  By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.



Type: Primary
Antigen: EPHB1 (EPH receptor B1)
Clonality: monoclonal
Clone: EPR6457
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR