Drukuj

Anti-S100P Rabbit Monoclonal Antibody [clone: EPR6142]

Dostawca: Abcam
AB124743-100 AB124743-1ML
ABCAAB124743-100EA 2860 PLN
ABCAAB124743-100 ABCAAB124743-1ML
Anti-S100P Rabbit Monoclonal Antibody [clone: EPR6142]
Przeciwciała
Rabbit monoclonal [EPR6142] to S100P.

Uwaga: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: S100P
Clonality: Monoclonal
Clone: EPR6142
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR