Drukuj

Anti-Proteasome 20S alpha 5/PSMA5 Rabbit Monoclonal Antibody [clone: EPR5833]

Dostawca: Abcam
AB109533-100 AB109533-1ML
ABCAAB109533-100EA 3110 PLN
ABCAAB109533-100 ABCAAB109533-1ML
Anti-Proteasome 20S alpha 5/PSMA5 Rabbit Monoclonal Antibody [clone: EPR5833]
Przeciwciała
Type: Primary
Antigen: Proteasome 20S alpha 5
Clonality: monoclonal
Clone: EPR5833
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR