Drukuj
Anti-Acetyl Coenzyme A Carboxylase Rabbit Monoclonal Antibody [clone: EPR4971]
Dostawca: Abcam
AB109368-100 AB109368-20UL AB109368-1ML
ABCAAB109368-100EA 3090 PLN
ABCAAB109368-100 ABCAAB109368-20UL ABCAAB109368-1ML
Anti-Acetyl Coenzyme A Carboxylase Rabbit Monoclonal Antibody [clone: EPR4971]
Przeciwciała
Rabbit monoclonal [EPR4971] to Acetyl Coenzyme A Carboxylase.
Uwaga: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: Acetyl Coenzyme A Carboxylase
Clonality: Monoclonal
Clone: EPR4971
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human