Drukuj
Anti-CD74 Rabbit Monoclonal Antibody [clone: EPR4064]
Dostawca: Abcam
AB108393-100 AB108393-1ML
ABCAAB108393-100EA 3090 PLN
ABCAAB108393-100 ABCAAB108393-1ML
Anti-CD74 Rabbit Monoclonal Antibody [clone: EPR4064]
Przeciwciała
Rabbit monoclonal [EPR4064] to CD74.
Uwaga: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.
Type: Primary
Antigen: CD74
Clonality: Monoclonal
Clone: EPR4064
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human