Drukuj

Anti-Calsequestrin 2 Rabbit Monoclonal Antibody [clone: EPR4228]

Dostawca: Abcam
AB108381-100 AB108381-10 AB108381-1ML
ABCAAB108381-100EA 2700 PLN
ABCAAB108381-100 ABCAAB108381-10 ABCAAB108381-1ML
Anti-Calsequestrin 2 Rabbit Monoclonal Antibody [clone: EPR4228]
Przeciwciała
Rabbit monoclonal [EPR4228] to Calsequestrin 2.

Uwaga: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive license for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: Calsequestrin 2
Clonality: Monoclonal
Clone: EPR4228
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR