Drukuj

Anti-CDKA1 / DOC1 Rabbit Monoclonal Antibody [clone: Clone-R76]

Dostawca: Abcam
AB108290-100 AB108290-1ML
ABCAAB108290-100EA 2750 PLN
ABCAAB108290-100 ABCAAB108290-1ML
Anti-CDKA1 / DOC1 Rabbit Monoclonal Antibody [clone: Clone-R76]
Przeciwciała
Rabbit monoclonal [Clone-R76] to CDKA1 / DOC1.

Uwaga: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive license for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: CDKA1 / DOC1
Clonality: Monoclonal
Clone: Clone-R76
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR