Drukuj

Anti-CD44 Rabbit Monoclonal Antibody [clone: SP37]

Dostawca: Abcam
AB101531-100 AB101531-500 AB101531-1 AB101531-20UL
ABCAAB101531-100EA 2110 PLN
ABCAAB101531-100 ABCAAB101531-500 ABCAAB101531-1 ABCAAB101531-20UL
Anti-CD44 Rabbit Monoclonal Antibody [clone: SP37]
Przeciwciała
Rabbit monoclonal [SP37] to CD44.

Uwaga: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: CD44
Clonality: Monoclonal
Clone: SP37
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR