Drukuj

Anti-delta 1 Catenin/CAS Rabbit Monoclonal Antibody [clone: EPR357(2)]

Dostawca: Abcam
AB92514-100 AB92514-1ML
ABCAAB92514-100EA 3170 PLN
ABCAAB92514-100 ABCAAB92514-1ML
Anti-delta 1 Catenin/CAS Rabbit Monoclonal Antibody [clone: EPR357(2)]
Rabbit monoclonal [EPR357(2)] to delta 1 Catenin/CAS.

Uwaga: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: delta 1 Catenin/CAS
Clonality: Monoclonal
Clone: EPR357(2)
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse; Human
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR