Drukuj

Anti-Fibrinogen gamma chain Rabbit Monoclonal Antibody [clone: EPR3084]

Dostawca: Abcam
AB92481-100 AB92481-1ML
ABCAAB92481-100EA 3110 PLN
ABCAAB92481-100 ABCAAB92481-1ML
Anti-Fibrinogen gamma chain Rabbit Monoclonal Antibody [clone: EPR3084]
Przeciwciała
Uwaga:

All products are FOR RESEARCH USE ONLY.   PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES.  Customer is granted a limited use non-exclusive licence for its own internal research.  Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product.  By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.



Type: Primary
Antigen: FGG (fibrinogen gamma chain)
Clonality: monoclonal
Clone: EPR3084
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR