Drukuj

Anti-TRIM21/SS-A Rabbit Polyclonal Antibody

Dostawca: Abcam
AB91423-20 AB91423-100UL
ABCAAB91423-20EA 3110 PLN
ABCAAB91423-20 ABCAAB91423-100UL
Anti-TRIM21/SS-A Rabbit Polyclonal Antibody
Przeciwciała
Rabbit polyclonal to TRIM21/SS-A.

Uwaga: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: TRIM21/SS-A
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR