Drukuj

Anti-CD34 Rabbit Monoclonal Antibody [clone: EP373Y]

Dostawca: Abcam
AB81289-100 AB81289-1ML
ABCAAB81289-100EA 3450 PLN
ABCAAB81289-100 ABCAAB81289-1ML
Anti-CD34 Rabbit Monoclonal Antibody [clone: EP373Y]
Przeciwciała
Rabbit monoclonal [EP373Y] to CD34.

Uwaga: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: CD34
Clonality: Monoclonal
Clone: EP373Y
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse; Human; Rat
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR