Drukuj

Anti-alpha Actinin/ACTN1 Rabbit Monoclonal Antibody [clone: EP2527Y]

Dostawca: Abcam
AB68194-100 AB68194-1ML
ABCAAB68194-100EA 2110 PLN
ABCAAB68194-100 ABCAAB68194-1ML
Anti-alpha Actinin/ACTN1 Rabbit Monoclonal Antibody [clone: EP2527Y]
Przeciwciała
Rabbit monoclonal [EP2527Y] to alpha Actinin/ACTN1.

Uwaga: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: alpha Actinin/ACTN1
Clonality: Monoclonal
Clone: EP2527Y
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR