Drukuj

Anti-Acetyl Coenzyme A Carboxylase (phospho S79) Rabbit Monoclonal Antibody [clone: EP1885Y]

Dostawca: Abcam
AB68191-100 AB68191-20UL AB68191-1ML
ABCAAB68191-100EA 3090 PLN
ABCAAB68191-100 ABCAAB68191-20UL ABCAAB68191-1ML
Anti-Acetyl Coenzyme A Carboxylase (phospho S79) Rabbit Monoclonal Antibody [clone: EP1885Y]
Przeciwciała
Rabbit monoclonal [EP1885Y] to Acetyl Coenzyme A Carboxylase (phospho S79).

Uwaga: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: Acetyl Coenzyme A Carboxylase
Clonality: Monoclonal
Clone: EP1885Y
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR