Drukuj

Anti-GSK3 (alpha + beta) Rabbit Monoclonal Antibody [clone: EP933Y]

Dostawca: Abcam
AB62368-100 AB62368-20UL AB62368-1ML
ABCAAB62368-100EA 3140 PLN
ABCAAB62368-100 ABCAAB62368-20UL ABCAAB62368-1ML
Anti-GSK3 (alpha + beta) Rabbit Monoclonal Antibody [clone: EP933Y]
Przeciwciała
Rabbit monoclonal [EP933Y] to GSK3 (alpha + beta).

Uwaga: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive license for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: GSK3 (alpha + beta)
Clonality: Monoclonal
Clone: EP933Y
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR