Drukuj

Anti-gamma Synuclein/SNCG Rabbit Monoclonal Antibody [clone: EP1539Y]

Dostawca: Abcam
AB52633-100 AB52633-1ML
ABCAAB52633-100EA 2810 PLN
ABCAAB52633-100 ABCAAB52633-1ML
Anti-gamma Synuclein/SNCG Rabbit Monoclonal Antibody [clone: EP1539Y]
Przeciwciała
Rabbit monoclonal [EP1539Y] to gamma Synuclein/SNCG.

Uwaga: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: gamma Synuclein/SNCG
Clonality: Monoclonal
Clone: EP1539Y
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR