Drukuj
Anti-Lactate Dehydrogenase Rabbit Monoclonal Antibody [clone: EP1566Y]
Dostawca: Abcam
AB52488-100 AB52488-1ML AB52488-20UL
ABCAAB52488-100EA 3860 PLN
ABCAAB52488-100 ABCAAB52488-1ML ABCAAB52488-20UL
Anti-Lactate Dehydrogenase Rabbit Monoclonal Antibody [clone: EP1566Y]
Przeciwciała
Rabbit monoclonal [EP1566Y] to Lactate Dehydrogenase.
Uwaga: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.
Type: Primary
Antigen: Lactate Dehydrogenase
Clonality: Monoclonal
Clone: EP1566Y
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse;Rat;Human