Drukuj

Anti-Huntingtin Rabbit Monoclonal Antibody [clone: EP867Y]

Dostawca: Abcam
AB45169-100 AB45169-1ML
ABCAAB45169-100EA 3670 PLN
ABCAAB45169-100 ABCAAB45169-1ML
Anti-Huntingtin Rabbit Monoclonal Antibody [clone: EP867Y]
Przeciwciała
Rabbit monoclonal [EP867Y] to Huntingtin.

Uwaga: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: Huntingtin
Clonality: Monoclonal
Clone: EP867Y
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse;Human
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR