Drukuj

Anti-RASA1 Rabbit Monoclonal Antibody [clone: EP536Y]

Dostawca: Abcam
AB40677-100 AB40677-1ML
ABCAAB40677-100EA 2970 PLN
ABCAAB40677-100 ABCAAB40677-1ML
Anti-RASA1 Rabbit Monoclonal Antibody [clone: EP536Y]
Przeciwciała
Rabbit monoclonal [EP536Y] to RASA1.

Uwaga: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: RASA1
Clonality: Monoclonal
Clone: EP536Y
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Rat, Human
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR