Drukuj

Anti-14-3-3 alpha + beta Rabbit Monoclonal Antibody [clone: Y62]

Dostawca: Abcam
AB32560-100 AB32560-1ML
ABCAAB32560-100EA 3140 PLN
ABCAAB32560-100 ABCAAB32560-1ML
Anti-14-3-3 alpha + beta Rabbit Monoclonal Antibody [clone: Y62]
Przeciwciała
Rabbit monoclonal [Y62] to 14-3-3 alpha + beta.

Uwaga: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: 14-3-3 alpha + beta
Clonality: Monoclonal
Clone: Y62
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR