Drukuj

Anti-CD33 Mouse Monoclonal Antibody [clone: WM53]

Dostawca: Abcam
AB30371-100 AB30371-1MG AB30371-20UG
ABCAAB30371-100EA 2840 PLN
ABCAAB30371-100 ABCAAB30371-1MG ABCAAB30371-20UG
Anti-CD33 Mouse Monoclonal Antibody [clone: WM53]
Przeciwciała
Mouse monoclonal [WM53] to CD33.

Uwaga: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: CD33
Clonality: Monoclonal
Clone: WM53
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR