Drukuj

Anti-NUCB1 Rabbit Polyclonal Antibody

Dostawca: Aviva Systems Biology
P102230_P050 P102230_T100
AVIVP102230_P050EA 2010 PLN
AVIVP102230_P050 AVIVP102230_T100
Anti-NUCB1 Rabbit Polyclonal Antibody
Przeciwciała
Type: Primary
Antigen: NUCB1 (nucleobindin 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR