Drukuj

Anti-TFDP1 Rabbit Polyclonal Antibody

Dostawca: Aviva Systems Biology
AVIVP100879_P050EA 1930 PLN
AVIVP100879_P050
Anti-TFDP1 Rabbit Polyclonal Antibody
Przeciwciała
Type: Primary
Antigen: TFDP1 (transcription factor Dp-1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity:
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR