Drukuj

Anti-TCF3 Rabbit Polyclonal Antibody

Dostawca: Aviva Systems Biology
AVIVP100871_T100EA 1680 PLN
AVIVP100871_T100
Anti-TCF3 Rabbit Polyclonal Antibody
Przeciwciała
Type: Primary
Antigen: TCF3 (transcription factor 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Dog, Guinea pig, Human, Mouse, Pig, Rat
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR