Drukuj

Anti-DPH7 Rabbit Polyclonal Antibody

Dostawca: Aviva Systems Biology
AVIVARP69141_P050EA 1930 PLN
AVIVARP69141_P050
Anti-DPH7 Rabbit Polyclonal Antibody
Przeciwciała
Type: Primary
Antigen: DPH7 (diphthamide biosynthesis 7)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR