Drukuj

Anti-TAP1 Rabbit Polyclonal Antibody

Dostawca: Aviva Systems Biology
AVIVARP43628_P050EA 1930 PLN
AVIVARP43628_P050
Anti-TAP1 Rabbit Polyclonal Antibody
Przeciwciała
Type: Primary
Antigen: TAP1 (transporter 1, ATP-binding cassette, sub-family B (MDR/TAP))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Bovine, Guinea pig, Human, Mouse, Pig, Rat
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR