Drukuj

Anti-ERI1 Rabbit Polyclonal Antibody

Dostawca: Aviva Systems Biology
AVIVARP41158_T100EA 1680 PLN
AVIVARP41158_T100
Anti-ERI1 Rabbit Polyclonal Antibody
Przeciwciała
Type: Primary
Antigen: ERI1 (exoribonuclease 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Bovine
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR