Drukuj

Anti-DIS3L2 Rabbit Polyclonal Antibody

Dostawca: Aviva Systems Biology
AVIVARP41145_T100EA 1680 PLN
AVIVARP41145_T100
Anti-DIS3L2 Rabbit Polyclonal Antibody
Przeciwciała
Type: Primary
Antigen: DIS3L2 (DIS3 like 3'-5' exoribonuclease 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Bovine, Dog, Human, Mouse, Pig, Rat
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR