Drukuj

Anti-RBM18 Rabbit Polyclonal Antibody

Dostawca: Aviva Systems Biology
AVIVARP41071_P050EA 1930 PLN
AVIVARP41071_P050
Anti-RBM18 Rabbit Polyclonal Antibody
Przeciwciała
Type: Primary
Antigen: RBM18 (RNA binding motif protein 18)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Bovine, Dog, Human, Mouse, Pig, Rat
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR