Drukuj

Anti-RBM3 Rabbit Polyclonal Antibody

Dostawca: Aviva Systems Biology
AVIVARP40309_P050EA 1930 PLN
AVIVARP40309_P050
Anti-RBM3 Rabbit Polyclonal Antibody
Przeciwciała
Type: Primary
Antigen: RBM3 (RNA binding motif (RNP1, RRM) protein 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Bovine, Dog, Guinea pig, Human, Mouse, Rat
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR