Drukuj

Anti-SOX7 Rabbit Polyclonal Antibody

Dostawca: Aviva Systems Biology
AVIVARP37139_T100EA 1680 PLN
AVIVARP37139_T100
Anti-SOX7 Rabbit Polyclonal Antibody
Przeciwciała
Type: Primary
Antigen: SOX7 (SRY (sex determining region Y)-box 7)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Bovine, Human, Mouse, Rat
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR