Drukuj

Anti-KLHL36 Rabbit Polyclonal Antibody

Dostawca: Aviva Systems Biology
AVIVARP34615_P050EA 1930 PLN
AVIVARP34615_P050
Anti-KLHL36 Rabbit Polyclonal Antibody
Przeciwciała
Type: Primary
Antigen: KLHL36 (kelch-like family member 36)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Bovine, Human, Mouse, Rat
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR