Drukuj

Anti-RASSF7 Rabbit Polyclonal Antibody

Dostawca: Aviva Systems Biology
AVIVARP34390_T100EA 1680 PLN
AVIVARP34390_T100
Anti-RASSF7 Rabbit Polyclonal Antibody
Przeciwciała
Type: Primary
Antigen: RASSF7 (Ras association (RalGDS/AF-6) domain family (N-terminal) member 7)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Bovine, Dog, Human, Mouse, Pig, Rat
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR