Drukuj
Anti-AIRE Rabbit Polyclonal Antibody
Dostawca: Aviva Systems Biology
ARP32649_P050 ARP32649_T100
AVIVARP32649_P050EA 2010 PLN
AVIVARP32649_P050 AVIVARP32649_T100
Anti-AIRE Rabbit Polyclonal Antibody
Przeciwciała
Type: Primary
Antigen: AIRE (autoimmune regulator)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity:
Zobacz ceny