Drukuj

Anti-GTF2I Rabbit Polyclonal Antibody

Dostawca: Aviva Systems Biology
ARP31910_P050 ARP31910_T100
AVIVARP31910_P050EA 2010 PLN
AVIVARP31910_P050 AVIVARP31910_T100
Anti-GTF2I Rabbit Polyclonal Antibody
Przeciwciała
Type: Primary
Antigen: GTF2I (general transcription factor IIi)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity:
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR