Drukuj
Płytki do PCR, 96-dołkowe, z niską ramką
Dostawca: Avantor
Peqlab
732-3201EA 691 PLN
732-3201 732-3227
Płytki do PCR, 96-dołkowe, z niską ramką
Mikropłytki Płytki do PCR
PP. Compatible with all major thermal cyclers.
  • Greater rigidity than non-skirted plates and provides space for barcoding
  • Plates with segmented design can be cut into 24 and 48 well sections
  • Ultra-thin for effective heat transfer and minimum condensation
  • Cut away corner for easy orientation
  • Certified free from RNase, DNase and human DNA